Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCTAIRE1 Polyclonal Antibody

Catalog No. PIPA529418
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529418 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529418 Supplier Invitrogen™ Supplier No. PA529418

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, HeLa, HepG2, Neuro2A, GL261. Predicted reactivity: Mouse (98%), Rat (98%), Xenopus laevis (86%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

CDK16 (PCTK1)/cyclin Y belongs to the cdc2/cdkx subfamily of the ser/thr family of protein kinases. CDK16 plays a role in signal transduction cascades in terminally differentiated cells, in exocytosis, and in transport of secretory cargo from the endoplasmic reticulum. CDK16 is ubiquitously expressed with the highest levels detected in the brain and testis. CDK16 also contained the gene coding for ubiquitin-activating enzyme UBE1 that previously shown to mapped to Xp11.3.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCTAIRE1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene CDK16
Gene Accession No. Q00536, Q04735
Gene Alias Cdk16; cdk16.L; cdk16.S; Cell division protein kinase 16; Crk5; cyclin dependent kinase 16; cyclin-dependent kinase 16; cyclin-dependent kinase 16 L homeolog; cyclin-dependent kinase 16 S homeolog; FLJ16665; PCTAIRE; PCTAIRE protein kinase 1; PCTAIRE1; PCTAIRE-1 protein kinase, alternatively spliced; PCTAIRE-motif protein kinase 1; pctgaire; pctk1; PCTK1 protein; Serine/threonine-protein kinase PCTAIRE-1; testis secretory sperm-binding protein Li 224n
Gene Symbols CDK16
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PCTAIRE1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18555, 5127
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.