Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCTAIRE2 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP142688PCP
Description
PCTAIRE2 Polyclonal antibody specifically detects PCTAIRE2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation (Negative)Specifications
PCTAIRE2 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoprecipitation (Negative) | |
Cell division protein kinase 17, cyclin-dependent kinase 17, EC 2.7.11, EC 2.7.11.22, PCTAIRE protein kinase 2, PCTAIRE2, PCTAIRE-motif protein kinase 2, PCTK2, protein kinase cdc2-related PCTAIRE-2, Serine/threonine-protein kinase PCTAIRE-2 | |
The immunogen for this product maps to a region between residue 1 and 50 of human PCTAIRE protein kinase 2 using the numbering given in entry NP_002586.2 (GeneID 5128). | |
0.1 mL | |
Cell Cycle and Replication | |
5128 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry | |
PerCP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction