Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCTAIRE2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553301
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553301 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553301 Supplier Invitrogen™ Supplier No. PA553301
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RRLSLTLRGS QTIDESLSEL AEQMTIEENS SKDNEPIVKN GRPPTSHSMH SFLHQYTGSF KKPPLRRPHS VIGGSLGSFM AMPRNGSRLD IVHENLKMGS DGESDQASGT SSDEVQSPTG VCLRNRI Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 99%.

CDK17/cyclin Y serine/threonine kinase or cyclin-dependent kinase 17 belongs to the cdc2/cdkx subfamily of kinases. CDK17/cyclin Y has similarity to a rat protein that is thought to play a role in terminally differentiated neurons. The identification of a large family of cdc2-related kinases opens the possibility of combinatorial regulation of the cell cycle together with the emerging large family of cyclins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCTAIRE2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CDK17
Gene Accession No. Q00537
Gene Alias 6430598J10Rik; CDK17; Cell division protein kinase 17; cyclin dependent kinase 17; cyclin-dependent kinase 17; PCTAIRE protein kinase 2; PCTAIRE2; PCTAIRE-motif protein kinase 2; PCTK2; protein kinase cdc2-related PCTAIRE-2; serine/threonine-protein kinase PCTAIRE-2
Gene Symbols CDK17
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PCTAIRE2. Recombinant protein control fragment (Product #RP-91421).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5128
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.