Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCTAIRE3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567852
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567852 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567852 Supplier Invitrogen™ Supplier No. PA567852
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Mouse and Rat samples.

May play a role in signal transduction cascades in terminally differentiated cells. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCTAIRE3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Cdk18
Gene Accession No. O35832, Q04899, Q07002
Gene Alias AA682070; Cdk18; Cell division protein kinase 18; Ctk18; cyclin dependent kinase 18; cyclin-dependent kinase 18; PCTAIRE; PCTAIRE protein kinase 3; Pctaire3; PCTAIRE-motif protein kinase 3; Pctk3; Serine/threonine-protein kinase PCTAIRE-3
Gene Symbols Cdk18
Host Species Rabbit
Immunogen Synthesized peptide derived from human PCTAIRE3.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18557, 289019, 5129
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.