Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCYOX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531528
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531528 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531528 Supplier Invitrogen™ Supplier No. PA531528
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A431. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Prenylcysteine is released during the degradation of prenylated proteins. PCYOX1 catalyzes the degradation of prenylcysteine to yield free cysteines and a hydrophobic isoprenoid product (Tschantz et al., 1999 ).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCYOX1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene PCYOX1
Gene Accession No. Q9UHG3
Gene Alias 1200015P13Rik; AI115532; AI448340; chloride ion pump-associated 55 kDa protein; Clp55; KIAA0908; PCL1; Pcly; PCYOX1; Prenylcysteine lyase; prenylcysteine oxidase; Prenylcysteine oxidase 1; prenylcysteine oxidase; prenylcysteine oxidase 1; prenylcysteine oxidase-like; UNQ597/PRO1183
Gene Symbols PCYOX1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PCYOX1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51449
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.