Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PCYOX1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121698
This item is not returnable.
View return policy
Description
PCYOX1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Prenylcysteine is released during the degradation of prenylated proteins. PCYOX1 catalyzes the degradation of prenylcysteine to yield free cysteines and a hydrophobic isoprenoid product (Tschantz et al., 1999 ).
Specifications
PCYOX1 | |
Polyclonal | |
Unconjugated | |
PCYOX1 | |
1200015P13Rik; AI115532; AI448340; chloride ion pump-associated 55 kDa protein; Clp55; KIAA0908; PCL1; Pcly; PCYOX1; Prenylcysteine lyase; prenylcysteine oxidase; Prenylcysteine oxidase 1; prenylcysteine oxidase; prenylcysteine oxidase 1; prenylcysteine oxidase-like; UNQ597/PRO1183 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
51449, 66881 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
Q9CQF9, Q9UHG3 | |
PCYOX1 | |
Recombinant protein Prenylcysteine Oxidase 1. The antigen corresponds to amino acid range 174-505 of the target protein. | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction