Learn More
Invitrogen™ PCYOX1 Polyclonal Antibody

Description
Peptide sequence: IFSQETLTKA QILKLFLSYD YAVKKPWLAY PHYKPPEKCP SIILHDRLYY Sequence homology: Cow: 83%; Dog: 86%; Horse: 86%; Human: 100%; Pig: 83%; Rabbit: 100%; Yeast: 75%.
Specifications
Specifications
| Antigen | PCYOX1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | PCYOX1 |
| Gene Accession No. | Q9UHG3 |
| Gene Alias | 1200015P13Rik; AI115532; AI448340; chloride ion pump-associated 55 kDa protein; Clp55; KIAA0908; PCL1; Pcly; PCYOX1; Prenylcysteine lyase; prenylcysteine oxidase; Prenylcysteine oxidase 1; prenylcysteine oxidase; prenylcysteine oxidase 1; prenylcysteine oxidase-like; UNQ597/PRO1183 |
| Gene Symbols | PCYOX1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.