Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PCYT1A Antibody (SR1699), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | PCYT1A |
---|---|
Clone | SR1699 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 |
Applications | Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation |
Classification | Monoclonal |
Description
PCYT1A Monoclonal antibody specifically detects PCYT1A in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, ImmunoprecipitationSpecifications
PCYT1A | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CCT A, CCT-alpha, choline-phosphate cytidylyltransferase A, CT, CT A, CTA, CTP:phosphocholine cytidylyltransferase A, CTPCTphosphate cytidylyltransferase 1, choline, alpha isoform, EC 2.7.7, EC 2.7.7.15, PCYT1CCTA, phosphate cytidylyltransferase 1, choline, alpha, Phosphorylcholine transferase A | |
A synthesized peptide derived from human PCYT1A (Uniprot #: P49585) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1699 | |
Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Cancer, Endocrinology, Neuroscience, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
5130 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title