Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCYT1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567924
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567924 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567924 Supplier Invitrogen™ Supplier No. PA567924
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PCYT1A Polyclonal Antibody for Western Blot, IHC

This gene belongs to the cytidylyltransferase family and is involved in the regulation of phosphatidylcholine biosynthesis. Mutations in this gene are associated with spondylometaphyseal dysplasia with cone-rod dystrophy. Alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCYT1A
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PCYT1A
Gene Accession No. P49585
Gene Alias CCT A; CCTA; Cctalpha; CCT-alpha; choline phosphate cytidylyltransferase 1 alpha; choline-phosphate cytidylyltransferase A; CT; CT A; CTA; CTalpha; CTP; CTP phosphocholine cytidylyl transferase; CTP:phosphocholine cytidylyl transferase; CTP:phosphocholine cytidylyltransferase A; CTP:phosphocholine cytidylyltransferase alpha; Ctpct; Cttalpha; PCYT1; PCYT1A; phosphate cytidylyltransferase 1, choline, alpha; phosphate cytidylyltransferase 1, choline, alpha isoform; phosphorylcholine transferase A; SMDCRD
Gene Symbols PCYT1A
Host Species Rabbit
Immunogen Synthesized peptide derived from human PCYT1A.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5130
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.