Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PCYT1A Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories

Catalog No. 501568039
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-8039 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-8039 Supplier Bethyl Laboratories, Inc Supplier No. A304559A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.

Controls phosphatidylcholine synthesis.

Specifications

Antigen PCYT1A
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene PCYT1A
Gene Accession No. P49585, P49586
Gene Alias CCT A, CCTA, CCT-alpha, CT, CT A, CTA, CTP:phosphocholine cytidylyltransferase A, CTPCT, PCYT1, phosphate cytidylyltransferase 1, choline, alpha isoform, phosphorylcholine transferase A, SMDCRD
Gene Symbols PCYT1A
Host Species Rabbit
Immunogen Between 317 and 367
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13026, 5130
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.