Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PCYT1B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114301
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114301 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114301 Supplier Invitrogen™ Supplier No. PA5114301
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SREFIGNFLE LFGPDGAWKQ MFQERSSRML QALSPKQSPV SSPTRSRSPS For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles.

The protein encoded by this gene belongs to the cytidylyltransferase family. It is involved in the regulation of phosphatidylcholine biosynthesis. Several alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PCYT1B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Pcyt1b
Gene Accession No. Q9Y5K3
Gene Alias AW045697; CCT B; CCTB; Cctbeta; CCT-beta; choline phosphate cytidylyltransferase 1 beta; choline-phosphate cytidylyltransferase B; CT B; CTB; CTP:phosphocholine cytidylyltransferase B; CTTbeta; PCYT1B; phosphate cytidylyltransferase 1, choline, beta; phosphate cytidylyltransferase 1, choline, beta isoform; phosphorylcholine transferase B
Gene Symbols Pcyt1b
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C terminal region of human PCYT1B (aa 282-331).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9468
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.