Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PD 0325901
GREENER_CHOICE

Catalog No. 419210
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-9210 10 mg
41-925-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 41-9210 Supplier Tocris Bioscience™ Supplier No. 4192/10
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Potent inhibitor of MEK1/2

PD 0325901 is a potent MEK1 and MEK2 inhibitor. PD 0325901 inhibits MEK activity in mouse colon 26 cells (IC50 = 0.33 nM). PD 0325901 inhibits the growth of melanoma cell lines in vitro and in vivo; induces G1-phase cell cycle arrest and apoptosis in a mouse xenograft model. Also inhibits production of proangiogenic cytokines such as VEGF. Enhances generation of induced pluripotent stem cells (iPSCs). Orally active.

For more information about how PD 0325901 may be used, see our protocols: Culturing Transcriptomically Distinct Pluripotent mESCs, Highly Efficient Generation of CiPSCs from MEFs, Accelerated Induction of Cortical Neurons from hiPSCs .

Chemical Identifiers

CAS 391210-10-9
Molecular Formula C16H14F3IN2O4

Specifications

Quantity 10 mg
Formula Weight 482.19
Purity >99%
Molecular Weight (g/mol) 482.198
Chemical Name or Material PD 0325901
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.