Learn More
Description
For more information about how PD 0325901 may be used, see our protocols: Culturing Transcriptomically Distinct Pluripotent mESCs, Highly Efficient Generation of CiPSCs from MEFs, Accelerated Induction of Cortical Neurons from hiPSCs .
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 482.19 |
| Purity | >99% |
| Molecular Weight (g/mol) | 482.198 |
| Chemical Name or Material | PD 0325901 |
Safety and Handling
| Recommended Storage | Store at -20°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.


