Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ PD 0332991 isethionate
GREENER_CHOICESDP

Catalog No. 478650 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-865-0 50 mg
47-861-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 47-865-0 Supplier R&D Systems™ Supplier No. 4786/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent, selective Cdk4/6 inhibitor; brain penetrant

Potent cyclin-dependent kinase 4 (cdk4) and cdk6 inhibitor (IC50 values are 11 nM and 15 nM respectively). Also displays activity at cdk2, cdk5 and cdk9 (IC50 values reported to be 1.6, 1.8 and 0.42-1.1 μM, respectively). Induces G1 cell cycle arrest and senescence in retinoblastoma protein (Rb)-proficient cell lines; blocks growth of intracranial glioblastoma multiforme xenografts in mice. Brain penetrant.

Chemical Identifiers

CAS 827022-33-3
Molecular Formula C26H35N7O6S

Specifications

Assay Percent Range >98%
Quantity 50 mg
Product Line Tocris
Formula Weight 573.66
Molecular Weight (g/mol) 573.669
Chemical Name or Material PD 0332991 isethionate
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.