Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    PD-1 Antibody (1015829), Allophycocyanin, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB10868A
                        
                            
                        
                        
                        
                            
                        
                        
                        
                        
                            
                                    
                                This item is not returnable.
                                View return policy
                                
                        
                    
                Description
PD-1 Monoclonal antibody specifically detects PD-1 in Human samples. It is validated for Flow CytometrySpecifications
| PD-1 | |
| Monoclonal | |
| APC | |
| CD279, CD279 antigen, hPD-1, PD1hPD-l, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2 | |
| Human embryonic kidney cell HEK293-derived human PD-1, Leu25-Thr168, Accession # Q15116 | |
| 0.1 mL | |
| Apoptosis, Protein Phosphatase, Stem Cell Markers | |
| 5133 | |
| Store at 4°C in the dark. | |
| IgG1 | 
| Flow Cytometry | |
| 1015829 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            