Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PD-1 Antibody (1026519) [HRP], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB10866H
Description
PD-1 Monoclonal antibody specifically detects PD-1 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
PD-1 | |
Monoclonal | |
HRP | |
PBS | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, CyTOF | |
1026519 | |
Western Blot, Flow Cytometry, CyTOF-ready | |
CD279, CD279 antigen, hPD-1, PD1hPD-l, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2 | |
Synthetic peptide containing human PD-1 | |
0.1 mL | |
Apoptosis, Protein Phosphatase, Stem Cell Markers | |
5133 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction