Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PD-1 Antibody (996221) [DyLight 594], Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB77381M
Description
PD-1 Monoclonal antibody specifically detects PD-1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
PD-1 | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Western Blot, Flow Cytometry, CyTOF | |
996221 | |
Western Blot, Flow Cytometry, CyTOF-ready | |
CD279, CD279 antigen, hPD-1, PD1hPD-l, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2 | |
Mouse myeloma cell line, NS0-derived mouse PD-1, Met1-Gln167, Accession # Q02242 | |
0.1 mL | |
Apoptosis, Protein Phosphatase, Stem Cell Markers | |
5133 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction