Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PD-1 Antibody (996221), PE/Atto594, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB77381PEATT594
Description
PD-1 Monoclonal antibody specifically detects PD-1 in Mouse samples. It is validated for Flow CytometrySpecifications
PD-1 | |
Monoclonal | |
PE/ATTO 594 | |
PBS | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry | |
996221 | |
Flow Cytometry | |
CD279, CD279 antigen, hPD-1, PD1hPD-l, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2 | |
Mouse myeloma cell line, NS0-derived mouse PD-1, Met1-Gln167, Accession # Q02242 | |
0.1 mL | |
Apoptosis, Phospho Specific, Protein Phosphatase, Stem Cell Markers | |
5133 | |
Store at 4C in the dark. Do not freeze. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction