Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PD-1 Antibody (CL3624), Novus Biologicals™

Mouse Monoclonal Antibody
$382.00 - $646.00
Specifications
Antigen | PD-1 |
---|---|
Clone | CL3624 |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
PD-1 Monoclonal specifically detects PD-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PD-1 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Human | |
5133 | |
This antibody was developed against a recombinant protein corresponding to amino acids: WFLDSPDRPWNPPTFSPALLVVTEGDNATFTCSFSNTSESFVLNWYRMSPSNQTDKLAAFPEDRSQPGQDCRFRVTQLPNGRDFHMSVVRARRNDSGTYLCGAIS | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
CL3624 | |
Monoclonal | |
Purified | |
Apoptosis, Protein Phosphatase, Stem Cell Markers | |
CD279, CD279 antigen, hPD-1, PD1hPD-l, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2 | |
PDCD1 | |
IgG1 | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title