Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PD-1 Antibody (Hu121), mFluor Violet 500 SE, Novus Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals FAB11380MFV500
Description
PD-1 Monoclonal antibody specifically detects PD-1 in Human samples. It is validated for Flow CytometrySpecifications
PD-1 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Human | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
Hu121 | |
Flow Cytometry | |
CD279, CD279 antigen, hPD-1, PD1hPD-l, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2 | |
Human PD-1 | |
0.1 mL | |
Apoptosis, Phospho Specific, Protein Phosphatase, Stem Cell Markers | |
5133 | |
Store at 4C in the dark. | |
IgG4 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction