Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PD-1 Antibody (J116) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP234498
Description
PD-1 Monoclonal specifically detects PD-1 in Human samples. It is validated for Flow Cytometry, Functional, CyTOF-ready.Specifications
PD-1 | |
Monoclonal | |
1 mg/mL | |
Flow Cytometry, Functional, CyTOF-ready | |
PDCD1 | |
The immunogen for this antibody was PD1. | |
0.1 mg | |
Primary | |
Human | |
IgG1 κ |
Flow Cytometry, Functional Assay, CyTOF | |
J116 | |
Unconjugated | |
CD279, CD279 antigen, hPD-1, PD1hPD-l, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2 | |
Mouse | |
Affinity Purified | |
RUO | |
5133 | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction