Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PD-1 Antibody (J43) - Azide and BSA Free, Novus Biologicals™

Armenian Hamster Monoclonal Antibody
$366.00
Specifications
Antigen | PD-1 |
---|---|
Clone | J43 |
Concentration | 1 mg/mL |
Dilution | Flow Cytometry, Functional, CyTOF-ready |
Applications | Flow Cytometry, Functional Assay, CyTOF |
Description
PD-1 Monoclonal specifically detects PD-1 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Functional, CyTOF-ready.Specifications
PD-1 | |
1 mg/mL | |
Flow Cytometry, Functional Assay, CyTOF | |
Unconjugated | |
RUO | |
CD279, CD279 antigen, hPD-1, PD1hPD-l, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2 | |
PDCD1 | |
IgG | |
Affinity Purified |
J43 | |
Flow Cytometry, Functional, CyTOF-ready | |
Monoclonal | |
Armenian Hamster | |
Mouse | |
5133 | |
The immunogen for this antibody was PD1. | |
Primary | |
Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title