Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PD-1 Antibody (J43), PE, Novus Biologicals™

Armenian Hamster Monoclonal Antibody
$366.00 - $499.50
Specifications
Antigen | PD-1 |
---|---|
Clone | J43 |
Concentration | 0.2 mg/mL |
Dilution | Flow Cytometry 1:10-1:1000 |
Applications | Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP14370005
![]() |
Novus Biologicals
NBP1437000.05MG |
0.05 mg |
Each for $366.00
|
|
|||||
NBP14370001
![]() |
Novus Biologicals
NBP1437000.1MG |
0.1 mg |
Each for $499.50
|
|
|||||
Description
PD-1 Monoclonal specifically detects PD-1 in Mouse samples. It is validated for Flow Cytometry.Specifications
PD-1 | |
0.2 mg/mL | |
Flow Cytometry | |
PE | |
RUO | |
Mouse | |
CD279, CD279 antigen, hPD-1, PD1hPD-l, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2 | |
PDCD1 | |
IgG | |
Affinity Purified |
J43 | |
Flow Cytometry 1:10-1:1000 | |
Monoclonal | |
Armenian Hamster | |
Apoptosis, Stem Cell Markers | |
Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide | |
5133 | |
The immunogen for this antibody was PD1. | |
Primary | |
Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title