Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PD-1 Antibody (PDCD1/1410R) - Azide and BSA Free, Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

$646.00

Specifications

Antigen PD-1
Clone PDCD1/1410R
Concentration 1 mg/mL
Dilution Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml, CyTOF-ready
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB254570100
SDP
View Documents
Novus Biologicals
NBP254570100UG
100 μg
Each for $646.00
Only null left
Add to Cart
 
NB254570200
SDP
View Documents
Novus Biologicals
NBP254570200UG
200 μg N/A N/A N/A
Description

Description

PD-1 Monoclonal specifically detects PD-1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

PD-1
1 mg/mL
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
Unconjugated
Rabbit
Apoptosis, Phospho Specific, Protein Phosphatase, Stem Cell Markers
PBS with No Preservative
5133
Human recombinant PDCD1 protein fragment (exact sequence is proprietary)
Primary
Store at -20 to -80C. Avoid freeze-thaw cycles.
PDCD1/1410R
Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml, CyTOF-ready
Monoclonal
Purified
RUO
Human
CD279, CD279 antigen, hPD-1, PD1hPD-l, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2
PDCD1
IgG
Protein A or G purified
55 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.