Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PD-1 Antibody (PDCD1/1410R), Novus Biologicals™

Rabbit Monoclonal Antibody
$304.50 - $548.00
Specifications
| Antigen | PD-1 |
|---|---|
| Clone | PDCD1/1410R |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml |
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB25320920
![]() |
Novus Biologicals
NBP25320920UG |
20 μg |
Each for $304.50
|
|
|||||
NB253209100
![]() |
Novus Biologicals
NBP253209100UG |
100 μg |
Each for $548.00
|
|
|||||
NB253209200
![]() |
Novus Biologicals
NBP253209200UG |
200 μg | N/A | N/A | N/A | ||||
Description
PD-1 Monoclonal specifically detects PD-1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| PD-1 | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Phospho Specific, Protein Phosphatase, Stem Cell Markers | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| 5133 | |
| Human recombinant PDCD1 protein fragment (exact sequence is proprietary) | |
| Primary | |
| Store at 4C. |
| PDCD1/1410R | |
| Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD279, CD279 antigen, hPD-1, PD1hPD-l, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2 | |
| PDCD1 | |
| IgG | |
| Protein A or G purified | |
| 55 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title