Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PD-1 Mouse anti-Human, Clone: EH33.1F9.G4.5, MilliporeSigma™

Catalog No. MABC1122100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC1122100 100 μg
MABC1122-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABC1122100 Supplier MilliporeSigma Supplier No. MABC1122100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-PD-1, clone EH33 is a mouse monoclonal antibody that detects Programmed cell death protein 1 (PD-1) in human cells and has been tested for use in Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PD-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone EH33.1F9.G4.5
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols PDCD1,PD1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to the extracellular domain of human PD-1 (as an Ig fusion protein).
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_005009.2
Test Specificity Clone EH33 specifically detects PD-1 in human cells. It targets an epitope with the extracellular domain.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.