Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ PD 150606
GREENER_CHOICESDP

Catalog No. 126950 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-695-0 50 mg
12-691-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 12-695-0 Supplier R&D Systems™ Supplier No. 1269/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Cell permeable calpain inhibitor

Selective, cell-permeable non-peptide calpain inhibitor (Ki values for ν and m-calpains are 0.21 and 0.37 μM respectively). Targets the calcium binding sites of calpain. Demonstrates high specificity for calpains relative to other proteases, and is a non-competitive inhibitor with respect to the substrate.

Chemical Identifiers

CAS 179528-45-1
Molecular Formula C9H7IO2S

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 306.12
Purity >95%
Molecular Weight (g/mol) 306.117
Chemical Name or Material PD 150606
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.