Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PD 158780
GREENER_CHOICE

Catalog No. 261510
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-151-0 10 mg
26-155-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 26-151-0 Supplier Tocris Bioscience™ Supplier No. 2615/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent ErbB receptor family inhibitor

Potent inhibitor of ErbB receptor family tyrosine kinases (IC50 values are 0.008, 49 and 52 nM for EGFR, ErbB2 and ErbB2/ErbB4 respectively) that does not inhibit FGF or PDGF-mediated tyrosine phosphorylation. Induces G1 cell cycle arrest in MCF10A cells and is antiproliferative in A431 human epidermal carcinoma cells.

Chemical Identifiers

CAS 171179-06-9
Molecular Formula C14H12BrN5

Specifications

Quantity 10 mg
Formula Weight 330.18
Purity >99%
Molecular Weight (g/mol) 330.19
Chemical Name or Material PD 158780
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.