Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PD 161570
GREENER_CHOICESDP

Catalog No. 372410R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-241-0R 10 mg
37-245-0R 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 37-241-0R Supplier Tocris Bioscience™ Supplier No. 3724/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective FGFR inhibitor

PD 161570 is a selective FGFR inhibitor (IC50 values are 40, 262 and 3700 nM for FGFR, PDGFR and EGFR respectively). Inhibits FGFR receptor phosphorylation (IC50 = 622 nM) and growth of A121 cells in vitro.

Specifications

Quantity 10 mg
Product Type PD 161570
Purity 0.97
Molecular Weight (g/mol) 532.51
Inhibitors PD 161570

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.