Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ PD 168077 maleate
GREENER_CHOICESDP

Catalog No. 106550 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-655-0 50 mg
10-651-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 10-655-0 Supplier R&D Systems™ Supplier No. 1065/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. High affinity, selective D4 agonist

A potent D4 dopamine receptor agonist (Ki = 8.7 nM) with > 400-fold selectivity over D2 and > 300-fold selectivity versus D3 subtypes respectively. Induces synaptic translocation of CaMK II to postsynaptic sites in cultured prefrontal cortical neurons. Centrally active in vivo.

Chemical Identifiers

CAS 630117-19-0
Molecular Formula C24H26N4O5

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 450.49
Purity 98%
Molecular Weight (g/mol) 450.495
Chemical Name or Material N-[[4-(2-Cyanophenyl)-1-piperazinyl]methyl]-3-methylbenzamide maleate
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.