Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ PD 173074
GREENER_CHOICESDP

Catalog No. 304450 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-445-0 50 mg
30-441-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 30-445-0 Supplier R&D Systems™ Supplier No. 3044/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. FGFR1 and -3 inhibitor

PD 173074 is a selective FGFR1 and FGFR3 inhibitor (IC50 values are 5, 21.5, ∼100, 17600 and 19800 nM for FGFR3, FGFR1, VEGFR2, PDGFR and c-Src respectively, and > 50000 nM for EGFR, InsR, MEK and PKC). Inhibits VEGF- and FGF-induced angiogenesis in the mouse cornea model of angiogenesis. Inhibits proliferation and differentiation of oligodendrocyte progenitors. Also promotes mESC self-renewal, facilitates the conversion of mouse epiblast stem cells to an earlier pluripotency state and inhibits differentiation of miPSCs to cardiomyocytes. Suppresses cell proliferation in cell lines expressing mutated FGFR3 protein. Blocks tumor growth in H510 and H69 SCLC xenograft models.

Chemical Identifiers

CAS 219580-11-7
Molecular Formula C28H41N7O3

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 523.67
Purity >98%
Molecular Weight (g/mol) 523.682
Chemical Name or Material PD 173074
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.