Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PD 176252
GSA_VAGREENER_CHOICESDP

Catalog No. 260210 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-021-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 26-021-0 Supplier Tocris Bioscience™ Supplier No. 2602/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

GRP (BB2) and NMB (BB1) receptor antagonist

Non-peptide gastrin-releasing peptide receptor (GRP-R, BB2) and neuromedin B receptor (NMB-R, BB1) antagonist (Ki values are 0.17 and 1.0 nM for BB1 and BB2 respectively). Inhibits proliferation of rat C6 glioma cells (IC50 = 2 μM) and inhibits NCI-H1299 xenograft proliferation in nude mice (IC50 = 5 μM).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 204067-01-6
Molecular Formula C32H36N6O5

Specifications

Quantity 10 mg
Formula Weight 584.67
Purity >98%
Molecular Weight (g/mol) 584.677
Chemical Name or Material PD 176252
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.