Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PD 334581
GREENER_CHOICESDP

Catalog No. 482410 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-241-0 10 mg
48-245-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 48-241-0 Supplier Tocris Bioscience™ Supplier No. 4824/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

MEK1 inhibitor

Inhibitor of MEK1. Analog of PD 184352 (Cat. No. 4237).

Chemical Identifiers

CAS 548756-68-9
Molecular Formula C20H19F3IN5O2

Specifications

Quantity 10 mg
Formula Weight 545.3
Purity >98%
Molecular Weight (g/mol) 545.305
Chemical Name or Material PD 334581
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.