Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ PD 407824
GREENER_CHOICESDP

Catalog No. 269410 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-941-0 10 mg
26-941 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 26-941-0 Supplier R&D Systems™ Supplier No. 2694/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective inhibitor of Chk1 and Wee1

Selective inhibitor of checkpoint kinases Chk1 and Wee1 (IC50 values are 47 and 97 nM respectively). Displays selectivity over a range of other protein kinases; IC50 values are 3.4, 3.75, > 5, > 50, > 50 and > 50 μM for PKC, CDK4, other CDKs, c-Src, PDGFR and FGFR respectively.

Chemical Identifiers

CAS 622864-54-4
Molecular Formula C20H12N2O3

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 328.32
Purity >98%
Molecular Weight (g/mol) 328.33
Chemical Name or Material PD 407824
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.