Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PD 81723
GREENER_CHOICE

Catalog No. 136310
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-631-0 10 mg
13-635-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 13-631-0 Supplier Tocris Bioscience™ Supplier No. 1363/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Allosteric potentiator of A1 receptors

Allosteric potentiator at the adenosine A1 receptor; acts via agonist-dependent and -independent mechanisms. Enhances agonist affinity for, and increased t½ of dissociation from, the receptor. Also inhibits basal and forskolin-stimulated adenylyl cyclase (AC) activity in A1 receptors expressed in CHO cells, possibly via direct potentiation of constitutive receptor activity or by direct inhibition of AC. Active in vivo.

Chemical Identifiers

CAS 132861-87-1
Molecular Formula C14H12F3NOS

Specifications

Quantity 10 mg
Formula Weight 299.31
Purity >99%
Molecular Weight (g/mol) 299.311
Chemical Name or Material PD 81723
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.