Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PD 90780

Catalog No. 510150
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-015-0 50 mg
51-011-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 51-015-0 Supplier Tocris Bioscience™ Supplier No. 5101/50

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Inhibits NGF binding to p75NTR

PD 90780 is a a nonpeptide inhibitor of NGF binding to p75NTR (IC50 ∽ 1 μM in CHO cells); binds NGF, not p75NTR. Affinity maybe altered by the presence of TrkA receptors. Neuroprotective.

Specifications

Purity >98%
Quantity 50 mg
Product Type PD 90780
Molecular Weight (g/mol) 362.34
Inhibitors PD 90780

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.