Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PD-ECGF/Thymidine Phosphorylase Antibody (TYMP/2890R) - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308402
Description
PD-ECGF/Thymidine Phosphorylase Monoclonal specifically detects PD-ECGF/Thymidine Phosphorylase in Human, Mouse, Rat samples. It is validated for Immunohistochemistry-Paraffin.Specifications
PD-ECGF/Thymidine Phosphorylase | |
Monoclonal | |
1 mg/mL | |
Immunohistochemistry-Paraffin : 1-2 ug/mL | |
EC 2.4.2, EC 2.4.2.4, ECGF, ECGF1TP, endothelial cell growth factor 1 (platelet-derived), Gliostatin, hPD-ECGF, MEDPS1, MNGIE, MTDPS1, PDECGF, PD-ECGF, Platelet-derived endothelial cell growth factor, TdRPase, thymidine phosphorylase | |
Recombinant full-length human PD-ECGF/Thymidine Phosphorylase (TP/ PD-ECGF) protein | |
100 μg | |
Angiogenesis | |
1890 | |
Store at -20 to -80C. Avoid freeze-thaw cycles. | |
IgG |
Immunohistochemistry (Paraffin) | |
TYMP/2890R | |
Unconjugated | |
10mM PBS | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction