Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PD-ECGF/Thymidine Phosphorylase Antibody (TYMP/2890R), HRP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308402H
Description
PD-ECGF/Thymidine Phosphorylase Monoclonal antibody specifically detects PD-ECGF/Thymidine Phosphorylase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
PD-ECGF/Thymidine Phosphorylase | |
Monoclonal | |
HRP | |
EC 2.4.2, EC 2.4.2.4, ECGF, ECGF1TP, endothelial cell growth factor 1 (platelet-derived), Gliostatin, hPD-ECGF, MEDPS1, MNGIE, MTDPS1, PDECGF, PD-ECGF, Platelet-derived endothelial cell growth factor, TdRPase, thymidine phosphorylase | |
Recombinant full-length human PD-ECGF/Thymidine Phosphorylase protein (Uniprot: P19971) | |
100 μg | |
Angiogenesis | |
1890 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry (Paraffin) | |
TYMP/2890R | |
PBS | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction