Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PD-L1 Antibody (2096A), Alexa Fluor™ 594, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals MAB90782AF594
Description
PD-L1 Monoclonal specifically detects PD-L1 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
PD-L1 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
CD274 | |
Mouse myeloma cell line NS0-derived recombinant mouse B7-H1/PD-L1, Met1-Thr239, Accession No. Q9EP73 | |
Autoimmune Diseases, Cancer, Cellular Markers, Checkpoint signaling, Immune System Diseases, Immunology, Signal Transduction, Tumor Biomarkers, Tumor Suppressors | |
29126 | |
Store at 4°C in the dark. |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
2096A | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
Avelumab, B7-H, B7-H1, B7H1PDCD1L1, CD274 antigenMGC142294, CD274 molecule, PDCD1LG1MGC142296, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1 | |
Rabbit | |
0.1 mL | |
Primary | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction