Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PD-L1 Antibody (Hu125), PE/Cy7, Novus Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals FAB10355PECY7
Description
PD-L1 Monoclonal antibody specifically detects PD-L1 in Human samples. It is validated for Flow CytometrySpecifications
PD-L1 | |
Monoclonal | |
PE-Cyanine7 | |
PBS | |
Human | |
Protein A or G purified from cell culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
Hu125 | |
Flow Cytometry | |
Avelumab, B7-H, B7-H1, B7H1PDCD1L1, CD274 antigenMGC142294, CD274 molecule, PDCD1LG1MGC142296, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1 | |
Human PD-L1/B7-H1, | |
0.1 mL | |
Autoimmune Diseases, Cancer, Cellular Markers, Checkpoint signaling, Immune System Diseases, Immunology, Signal Transduction, Tumor Biomarkers, Tumor Suppressors | |
29126 | |
Store at 4C in the dark. Do not freeze. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction