Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PD-L1 Antibody (MIH5), Alexa Fluor™ 594, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP143262AF594
Description
PD-L1 Monoclonal specifically detects PD-L1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
PD-L1 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
CD274 | |
The immunogen for this antibody was B7H1. | |
Autoimmune Diseases, Cancer, Cellular Markers, Checkpoint signaling, Immune System Diseases, Immunology, Signal Transduction, Tumor Biomarkers, Tumor Suppressors | |
29126 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
MIH5 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
Avelumab, B7-H, B7-H1, B7H1PDCD1L1, CD274 antigenMGC142294, CD274 molecule, PDCD1LG1MGC142296, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1 | |
Rat | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG2a λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction