Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PD-L1 Antibody (MIH5) - BSA Free, Novus Biologicals™
SDP

Rat Monoclonal Antibody has been used in 5 publications

$192.47 - $666.00

Specifications

Antigen PD-L1/B7-H1
Clone MIH5
Concentration 0.5 mg/mL
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen 1:10-1:500
Applications Immunohistochemistry, Immunohistochemistry (Frozen), Flow Cytometry, Western Blot, Immunofluorescence, Immunocytochemistry
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018526
SDP
View Documents
Novus Biologicals
NBP14326225UG
25 μg
Each for $192.47
Only null left
Add to Cart
 
NBP14326201
SDP
View Documents
Novus Biologicals
NBP1432620.1MG
0.1 mg
Each for $493.10
Only null left
Add to Cart
 
NBP1432625
SDP
View Documents
Novus Biologicals
NBP1432620.5MG
0.5 mg
Each for $666.00
Only null left
Add to Cart
 
Description

Description

PD-L1 Monoclonal specifically detects PD-L1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, In vivo assay.
Specifications

Specifications

PD-L1/B7-H1
0.5 mg/mL
Immunohistochemistry, Immunohistochemistry (Frozen), Flow Cytometry, Western Blot, Immunofluorescence, Immunocytochemistry
Unconjugated
RUO
Human, Mouse
anti-PD-L1, B7-H, B7-H1, B7H1PDCD1L1, CD274 antigenMGC142294, CD274 molecule, PDCD1LG1MGC142296, PD-L1, PD-L1 blocking, PD-L1 ihc, PD-L1 immunohistochemistry, PD-L1 western blot, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1
CD274
IgG2a λ
Affinity Purified
MIH5
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen 1:10-1:500
Monoclonal
Rat
Signal Transduction
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
29126
The immunogen for this antibody was B7H1.
Primary
Store at 4°C. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.