Learn More
PD-L1/CD274 (C-terminal) Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | PD-L1/CD274 (C-terminal) |
---|---|
Concentration | 233 μg/mL |
Applications | Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-223-8617
![]() ![]() |
Proteintech Group Inc
280761AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The PD-L1/CD274 (C-terminal) antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PD-L1/CD274 (C-terminal). This antibody recognizes human, mouse, rat antigen. The PD-L1/CD274 (C-terminal) antibody has been validated for the following applications: IF, IHC, IP, WB, ELISA analysis.
Specifications
PD-L1/CD274 (C-terminal) | |
Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, ELISA | |
Unconjugated | |
Human, Rat, Mouse | |
IgG |
233 μg/mL | |
Polyclonal | |
Rabbit | |
29126 | |
Primary |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.