Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PD-L1 (CD274) Mouse anti-Human, Alexa Fluor™ 647, Clone: IPI652.rMAb (also known as IPI652.rMAb), BD Phrmingen™

Catalog No. BDB568925 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB568925 25 Tests
BDB568924 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB568925 Supplier BD Biosciences Supplier No. 568925
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PD-L1 (CD274) Monoclonal antibody specifically detects PD-L1 (CD274) in Human samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen PD-L1 (CD274)
Applications Flow Cytometry
Classification Monoclonal
Clone IPI652.rMAb (also known as IPI652.rMAb)
Conjugate Alexa Fluor 647
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias B7H1; PD-L1; PDCD1L1; PDCD1LG1; PDL1; hPD-L1
Host Species Mouse
Immunogen Human PD-L1
Purification Method Affinity Chromatography
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29126
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.