Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PD-L1 (CD274) Mouse anti-Human, APC, Clone: 29E.2A3, BD Pharmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 568315
This item is not returnable.
View return policy
Description
PD-L1 (CD274) Monoclonal antibody specifically detects PD-L1 (CD274) in Human samples. It is validated for Flow cytometry (Routinely Tested)
Specifications
PD-L1 (CD274) | |
Monoclonal | |
APC | |
B7H1; PD-L1; PDCD1L1; PDCD1LG1; PDL1; hPD-L1 | |
Human PD-L1 | |
100 Tests | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
29E.2A3 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
29126 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction