Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PD-L1 (CD274) Mouse anti-Human, Clone: 29E.2A3, BD Pharmingen™

Catalog No. BDB568081 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB568081 0.5 mg
BDB568082 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. BDB568081 Supplier BD Biosciences Supplier No. 568081
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PD-L1 (CD274) Monoclonal antibody specifically detects PD-L1 (CD274) in Human samples. It is validated for Flow Cytometry, Functional Assay assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen PD-L1 (CD274)
Applications Flow Cytometry, Functional Assay
Classification Monoclonal
Clone 29E.2A3
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation No azide/low endotoxin: Aqueous buffered solution containing no preservative, sterile filtered(0.2μm pore size membrane). Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay.
Gene Alias B7H1; PD-L1; PDCD1L1; PDCD1LG1; PDL1; hPD-L1
Host Species Mouse
Immunogen Human PD-L1
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. This preparation contains no preservatives, thus it should be handled under aseptic conditions.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.