Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PD-L1 (CD274) Mouse anti-Human, PE, Clone: 29E.2A3, BD Pharmingen™
Mouse Monoclonal Antibody
$138.60 - $307.50
Specifications
| Antigen | PD-L1 (CD274) |
|---|---|
| Clone | 29E.2A3 |
| Dilution | 0.02% Sodium Azide |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
PD-L1 (CD274) Monoclonal antibody specifically detects PD-L1 (CD274) in Human samples. It is validated for Flow CytometrySpecifications
| PD-L1 (CD274) | |
| 0.02% Sodium Azide | |
| Monoclonal | |
| Liquid | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human PD-L1 | |
| Primary |
| 29E.2A3 | |
| Flow Cytometry | |
| PE | |
| Mouse | |
| Human | |
| B7H1; PD-L1; PDCD1L1; PDCD1LG1; PDL1; hPD-L1 | |
| IgG2b κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title