Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human PD-L1/B7-H1 (Research Grade Atezolizumab Biosimilar) Alexa Fluor™ 594-conjugated Antibody, R&D Systems™

Human Monoclonal Antibody
Supplier: R&D Systems FAB10348T100UG
This item is not returnable.
View return policy
Description
PD-L1 Monoclonal specifically detects PD-L1 in Human samples. It is validated for Flow Cytometry.Specifications
PD-L1 | |
Monoclonal | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
Human | |
Protein A or G purified from cell culture supernatant | |
RUO | |
29126 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG1 |
Flow Cytometry | |
Hu124 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Avelumab, B7-H, B7H1, B7-H1, B7H1PDCD1L1, CD274, CD274 antigenMGC142294, CD274 molecule, PDCD1L1, PDCD1LG1, PDCD1LG1MGC142296, PDL1, PD-L1, PD-L1B7 homolog 1, PDL1PDCD1 ligand 1, programmed cell death 1 ligand 1, Programmed death ligand 1 | |
Human PD-L1/B7-H1 | |
100 μg | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction