Learn More
PD-L1 Mouse anti-Human, Clone: OTI12G7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF810925

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
PD-L1 (Programmed death receptor ligand 1, B7-H1, CD274) is a 40 kDa transmembrane protein and B7 family member. PD-L1 has been shown to negatively regulate T-cell receptor (TCR) signaling. Upon ligating its receptor, PD-L1 has been reported to decrease TCR-mediated proliferation and cytokine production, and PD-L1 expression was found to be abundant on many murine and human cancers and could be further up-regulated upon IFN-gamma stimulation. Thus, PD-L1 might play an important role in tumor immune evasion. Together with B7.1, PD-L1 can inactivate T cells and this relationship is a focal point of cancer therapeutic treatment. At least two isoforms of PDL-1 are known to exist.Specifications
PD-L1 | |
Monoclonal | |
Unconjugated | |
CD274 | |
B7-H, B7H1, PD-L1, PDCD1L1, PDCD1LG1, PDL1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
29126 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI12G7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9NZQ7 | |
CD274 | |
Human recombit protein fragment corresponding to amino acids 19-290 of human CD274 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.