Learn More
Promega PD-L1 Negative Cells, 1 each
Catalog No. PRJ1191 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 Each
Catalog No. Quantity
PRJ1191 1 Each
1 options
Catalog No. PRJ1191 Supplier Promega Supplier No. J1191
PD-L1 Negative Cells are control cells for use with the PD-1/PD-L1 Blockade Bioassay.
  • One−time, ready−to−use control cells for PD−1/PD−L1 Blockade Bioassay
Specifications
Content And Storage BELOW -140°C
Quantity 1 Each